Anti-Serine/threonine-protein Kinase MAK-V Rabbit Polyclonal Antibody
Lieferant: US Biological
H7953-31D
USBIH7953-31DEA 633 EUR
USBIH7953-31D
Anti-Serine/threonine-protein Kinase MAK-V Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: Serine/threonine-protein Kinase MAK-V
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human