Anti-Hyaluronidase Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
H7981-05D
USBIH7981-05DEA 501 EUR
USBIH7981-05D
Anti-Hyaluronidase Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Hyaluronidase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR