Anti-Hydroxyguanosine 8 Mouse Monoclonal Antibody [clone: 9L175]

Lieferant: US Biological
H9076-01A
USBIH9076-01AEA 755 EUR
USBIH9076-01A
Anti-Hydroxyguanosine 8 Mouse Monoclonal Antibody [clone: 9L175]
Antikörper
Type: Primary
Antigen: Hydroxyguanosine 8
Clonality: monoclonal
Clone: 9L175
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR