Anti-Hydroxyguanosine 8 Goat Polyclonal Antibody

Lieferant: US Biological
H9076-03
USBIH9076-03EA 614 EUR
USBIH9076-03
Anti-Hydroxyguanosine 8 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:250000; Immunochemistry: 1:200

Type: Primary
Antigen: Hydroxyguanosine 8
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR