Anti-5-Hydoxy-Indole Acetic Acid Rabbit Polyclonal Antibody (Unconjugated)
Lieferant: US Biological
H9090-55
USBIH9090-55EA 739 EUR
USBIH9090-55
Anti-5-Hydoxy-Indole Acetic Acid Rabbit Polyclonal Antibody (Unconjugated)
Antikörper
Type: Primary
Antigen: 5-Hydoxy-Indole Acetic Acid
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen