Anti-ICOS Hamster Monoclonal Antibody [clone: 10B1220]

Lieferant: US Biological
I0649-95
USBII0649-95EA 575 EUR
USBII0649-95
Anti-ICOS Hamster Monoclonal Antibody [clone: 10B1220]
Antikörper
Recommended dilution: Flow Cytometry: 1:5-1:10;

Type: Primary
Antigen: ICOS (inducible T-cell co-stimulator)
Clonality: monoclonal
Clone: 10B1220
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR