Anti-CD79A Goat Polyclonal Antibody (Rhodamine)

Lieferant: US Biological
I1890-06A7
USBII1890-06A7EA 411 EUR
USBII1890-06A7
Anti-CD79A Goat Polyclonal Antibody (Rhodamine)
Antikörper
Type: Primary
Antigen: CD79A (CD79a molecule, immunoglobulin-associated alpha)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR