Anti-IgA Goat Polyclonal Antibody (Texas Red®)

Lieferant: US Biological
I1890-28M
USBII1890-28MEA 451 EUR
USBII1890-28M
Anti-IgA Goat Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgA, Human
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR