Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
I1903-13M
USBII1903-13MEA 694 EUR
USBII1903-13M
Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000 to 1:2000
Do not freeze!

Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR