Anti-IgG Chicken Polyclonal Antibody (Texas Red®)

Lieferant: US Biological
I1903-14F
USBII1903-14FEA 594 EUR
USBII1903-14F
Anti-IgG Chicken Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR