Anti-IgG, F Rabbit Polyclonal Antibody (Texas Red®)
Lieferant: US Biological
I1903-21Q
USBII1903-21QEA 489 EUR
USBII1903-21Q
Anti-IgG, F Rabbit Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG, F
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Horse