Anti-IgG Mouse Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: US Biological
I1903-30L
USBII1903-30LEA 669 EUR
USBII1903-30L
Anti-IgG Mouse Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR