Anti-IgG Rat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Lieferant: US Biological
I1903-30M
USBII1903-30MEA 669 EUR
USBII1903-30M
Anti-IgG Rat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Rabbit