Anti-IgG Sheep Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: US Biological
I1903-30N
USBII1903-30NEA 669 EUR
USBII1903-30N
Anti-IgG Sheep Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR