Anti-IgG Goat Polyclonal Antibody (Texas Red®)
Lieferant: US Biological
I1903-35Y
USBII1903-35YEA 556 EUR
USBII1903-35Y
Anti-IgG Goat Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human