Anti-IgG F Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
I1903-38W
USBII1903-38WEA 688 EUR
USBII1903-38W
Anti-IgG F Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:200 to 1:1000
Do not freeze!

Type: Secondary
Antigen: IgG F
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR