Anti-IgE Goat Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
I1903-50J
USBII1903-50JEA 586 EUR
USBII1903-50J
Anti-IgE Goat Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: Flow Cytometry: 1:20-1:2000

Type: Primary
Antigen: IgE
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR