Anti-IgG Goat Polyclonal Antibody

Lieferant: US Biological
I1903-50Q
USBII1903-50QEA 338 EUR
USBII1903-50Q
Anti-IgG Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR