Anti-IgG F(c) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Lieferant: US Biological
I1903-52U
USBII1903-52UEA 298 EUR
USBII1903-52U
Anti-IgG F(c) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Recommended dilution: Flow Cytometry: 1:20-1:2000
Type: Secondary
Antigen: IgG Fc
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human