Anti-IgM Rabbit Polyclonal Antibody (TRITC (Rhodamine))
Lieferant: US Biological
I1903-54Y
USBII1903-54YEA 387 EUR
USBII1903-54Y
Anti-IgM Rabbit Polyclonal Antibody (TRITC (Rhodamine))
Antikörper
Recommended dilution: Flow Cytometry: 1:20-1:2000
Type: Primary
Antigen: IgM (IgM)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human