Anti-IgG Mouse Monoclonal Antibody Gold [clone: 10g102]

Lieferant: US Biological
I1903-77C
USBII1903-77CEA 357 EUR
USBII1903-77C
Anti-IgG Mouse Monoclonal Antibody Gold [clone: 10g102]
Antikörper
Type: Primary
Antigen: IgG (IgG)
Clonality: monoclonal
Clone: 10g102
Conjugation: Gold
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR