Anti-IgG Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
I1903-88D2
USBII1903-88D2EA 377 EUR
USBII1903-88D2
Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR