Anti-IgG Donkey Polyclonal Antibody (Biotin)

Lieferant: US Biological
I1903-89K2
USBII1903-89K2EA 426 EUR
USBII1903-89K2
Anti-IgG Donkey Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR