Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)

Lieferant: US Biological
I1904-04F-500
USBII1904-04F-500EA 405 EUR
USBII1904-04F-500
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR