Anti-IgG, F Goat Polyclonal Antibody (Texas Red®)

Lieferant: US Biological
I1904-43D
USBII1904-43DEA 518 EUR
USBII1904-43D
Anti-IgG, F Goat Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG, F
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR