Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein))
Lieferant: US Biological
I1904-45M4
USBII1904-45M4EA 338 EUR
USBII1904-45M4
Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: Immunochemistry: 1:20-1:5000
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Goat