Anti-IgG, F Goat Polyclonal Antibody (Texas Red®)
Lieferant: US Biological
I1904-46Z
USBII1904-46ZEA 435 EUR
USBII1904-46Z
Anti-IgG, F Goat Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG, F
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit