Anti-IgG F Goat Polyclonal Antibody (Biotin)
Lieferant: US Biological
I1904-50S1
USBII1904-50S1EA 573 EUR
USBII1904-50S1
Anti-IgG F Goat Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:20000 to 1:100000
Type: Secondary
Antigen: IgG F
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Rat