Anti-IgG Goat Polyclonal Antibody (Biotin)

Lieferant: US Biological
I1904-53G2
USBII1904-53G2EA 318 EUR
USBII1904-53G2
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR