Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
I1904-61E
USBII1904-61EEA 688 EUR
USBII1904-61E
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:10000 to 1:50000

Type: Secondary
Antigen: IgG H+L
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR