Anti-IgG Donkey Polyclonal Antibody (Texas Red®)

Lieferant: US Biological
I1904-63D
USBII1904-63DEA 600 EUR
USBII1904-63D
Anti-IgG Donkey Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Donkey
Isotype:
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR