Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
I1904-67AB
USBII1904-67ABEA 512 EUR
USBII1904-67AB
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:500 to 1:2500
Do not freeze!

Type: Secondary
Antigen: IgG H+L
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR