Anti-IgG Goat Polyclonal Antibody ATTO 488

Lieferant: US Biological
I1904-71
USBII1904-71EA 444 EUR
USBII1904-71
Anti-IgG Goat Polyclonal Antibody ATTO 488
Antikörper
Recommended dilution: Flow Cytometry: 1:100; Immunochemistry: 1:100

Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR