Anti-IgG Goat Polyclonal Antibody ATTO 488
Lieferant: US Biological
I1904-71D
USBII1904-71DEA 460 EUR
USBII1904-71D
Anti-IgG Goat Polyclonal Antibody ATTO 488
Antikörper
Recommended dilution: Flow Cytometry: 1:100; Immunochemistry: 1:100
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat