Anti-IgG2 F(c) Mouse Monoclonal Antibody [clone: 8.F.176]

Lieferant: US Biological
I1904-83A2
USBII1904-83A2EA 979 EUR
USBII1904-83A2
Anti-IgG2 F(c) Mouse Monoclonal Antibody [clone: 8.F.176]
Antikörper
Type: Secondary
Antigen: IgG2, Fc
Clonality: monoclonal
Clone: 8.F.176
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR