Anti-IgM Goat Polyclonal Antibody (TRITC (Rhodamine))

Lieferant: US Biological
I1905-04E6
USBII1905-04E6EA 266 EUR
USBII1905-04E6
Anti-IgM Goat Polyclonal Antibody (TRITC (Rhodamine))
Antikörper
Type: Primary
Antigen: IgM (IgM)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR