Anti-Influenza B Mouse Monoclonal Antibody [clone: 10D271]
Lieferant: US Biological
I7651-05A
USBII7651-05AEA 551 EUR
USBII7651-05A
Anti-Influenza B Mouse Monoclonal Antibody [clone: 10D271]
Antikörper
Recommended dilution: Immunofluorescence: 1:10-1:50; Immunochemistry: 1:10-1:50
Type: Primary
Antigen: Influenza B
Clonality: monoclonal
Clone: 10D271
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen