Anti-Inosine Monophosphate Dehydrogenase 2 Goat Polyclonal Antibody

Lieferant: US Biological
I7654-95A
USBII7654-95AEA 601 EUR
USBII7654-95A
Anti-Inosine Monophosphate Dehydrogenase 2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:8000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: Inosine Monophosphate Dehydrogenase 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR