Anti-Inositol Polyphosphate 4-phosphatase Type II Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
I7655-55B-AP
USBII7655-55B-APEA 884 EUR
USBII7655-55B-AP
Anti-Inositol Polyphosphate 4-phosphatase Type II Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: Inositol Polyphosphate 4-phosphatase Type II
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR