Anti-Inositol Polyphosphate 4-phosphatase Type II Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
I7655-55B-APC
USBII7655-55B-APCEA 884 EUR
USBII7655-55B-APC
Anti-Inositol Polyphosphate 4-phosphatase Type II Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: Inositol Polyphosphate 4-phosphatase Type II
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human