Anti-Inositol 1,4,5-trisphosphate 3-kinase A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
I7655-65C-AP
USBII7655-65C-APEA 884 EUR
USBII7655-65C-AP
Anti-Inositol 1,4,5-trisphosphate 3-kinase A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: Inositol 1,4,5-trisphosphate 3-kinase A
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR