Anti-Inositol 1,4,5-trisphosphate 3-kinase A Rabbit Polyclonal Antibody

Lieferant: US Biological
I7655-65N
USBII7655-65NEA 649 EUR
USBII7655-65N
Anti-Inositol 1,4,5-trisphosphate 3-kinase A Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: Inositol 1,4,5-trisphosphate 3-kinase A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR