Anti-Insulin-like 3 Rabbit Polyclonal Antibody (Carboxyfluorescein)

Lieferant: US Biological
I7660-85K2
USBII7660-85K2EA 1160 EUR
USBII7660-85K2
Anti-Insulin-like 3 Rabbit Polyclonal Antibody (Carboxyfluorescein)
Antikörper
Type: Primary
Antigen: Insulin-like 3
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR