Anti-Integrin, beta1 Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6A267]

Lieferant: US Biological
I7661-38R4
USBII7661-38R4EA 681 EUR
USBII7661-38R4
Anti-Integrin, beta1 Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6A267]
Antikörper
Recommended dilution: Flow Cytometry:1:5;
Do not freeze!

Type: Primary
Antigen: Integrin, beta1
Clonality: monoclonal
Clone: 6A267
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR