Anti-Integrin beta-like 1 with EGF-like Repeat Domains Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
I7661-49A-BIOT
USBII7661-49A-BIOTEA 884 EUR
USBII7661-49A-BIOT
Anti-Integrin beta-like 1 with EGF-like Repeat Domains Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Integrin beta-like 1 with EGF-like Repeat Domains
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR