Anti-Integrin beta-like 1 with EGF-like Repeat Domains Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
I7661-49A-PE
USBII7661-49A-PEEA 884 EUR
USBII7661-49A-PE
Anti-Integrin beta-like 1 with EGF-like Repeat Domains Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: Integrin beta-like 1 with EGF-like Repeat Domains
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human