Anti-Interleukin 1 Receptor Type I Hamster Monoclonal Antibody [clone: JAMA-147]

Lieferant: US Biological
I7663-25D1
USBII7663-25D1EA 481 EUR
USBII7663-25D1
Anti-Interleukin 1 Receptor Type I Hamster Monoclonal Antibody [clone: JAMA-147]
Antikörper
Recommended dilution: Flow Cytometry: 1:25-1:100;

Type: Primary
Antigen: Interleukin 1 Receptor Type I
Clonality: monoclonal
Clone: JAMA-147
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR