Anti-Interleukin 1 Receptor Type I Hamster Monoclonal Antibody [clone: JAMA-147]
Lieferant: US Biological
I7663-25D1A
USBII7663-25D1AEA 478 EUR
USBII7663-25D1A
Anti-Interleukin 1 Receptor Type I Hamster Monoclonal Antibody [clone: JAMA-147]
Antikörper
Recommended dilution: Flow Cytometry: 1:25-1:100;
Type: Primary
Antigen: Interleukin 1 Receptor Type I
Clonality: monoclonal
Clone: JAMA-147
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse