Anti-Insulin-like Growth Factor-like 3 Mouse Monoclonal Antibody [clone: 9L838]

Lieferant: US Biological
I7667-05
USBII7667-05EA 789 EUR
USBII7667-05
Anti-Insulin-like Growth Factor-like 3 Mouse Monoclonal Antibody [clone: 9L838]
Antikörper
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 2?g/ml; Immunochemistry: 25 ?g/ml

Type: Primary
Antigen: Insulin-like Growth Factor-like 3
Clonality: monoclonal
Clone: 9L838
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR