Anti-Interleukin 6 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
I8428-37-50
USBII8428-37-50EA 425 EUR
USBII8428-37-50
Anti-Interleukin 6 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Interleukin 6
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR